Isophthaloyl dichloride ≥99%

Synonyms: Isophthaloyl chloride

I19403-100G I19403-500G
220192-626EA 57.4 USD
220192-626 220192-627
Isophthaloyl dichloride ≥99%
Isophthaloyl dichloride
Formula: C₈H₄Cl₂O₂
MW: 203.02 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 42 - 45 °C
Density: 1.388 g/cm³ (17 °C)
MDL Number: MFCD00000678
CAS Number: 99-63-8
EINECS: 202-774-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR