Isophthalic acid ≥99%

I19209-2KG I19209-500G
220192-624EA 102.31 USD
220192-624 220192-625
Isophthalic acid ≥99%
Isophthalic acid
Formula: C₈H₆O₄
MW: 166.13 g/mol
Boiling Pt: 119 - 121 °C (6 -rr)
Melting Pt: 345 - 348 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002516
CAS Number: 121-91-5
EINECS: 204-506-4
Merck Index: 13,05216

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR