Isophorone ≥97%

Warning

Synonyms: 3,5,5-Trimethyl-2-cyclohexen-1-one, 3,5,5-Trimethylcyclohex-2-enone

I18709-1L I18709-5ML I18709-100ML
220192-608EA 65.69 USD
220192-608 220192-609 220192-607
Isophorone ≥97%
Isophorone
Formula: C₉H₁₄O
MW: 138.21 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: -8 °C
Density: 0.922 g/cm³ (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00001584
CAS Number: 78-59-1
EINECS: 201-126-0
Merck Index: 13,05215

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR