Isobutyric acid ≥99%

Warning

Synonyms: 2-Methylpropionic acid, 2-Methylpropanoic acid

I1754-1L I1754-500ML I1754-100ML
220192-150EA 172.31 USD
220192-150 220192-151 220192-149
Isobutyric acid ≥99%
Isobutyric acid
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: -45 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00002658
CAS Number: 79-31-2
EINECS: 201-195-7
UN: 2529
ADR: 3,III
Merck Index: 13,05172

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR