Isonicotinic acid

Warning

Synonyms: Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid

I17508-5G I17508-500G I17508-100G
220192-148EA 28.54 USD
220192-148 220192-147 220192-146
Isonicotinic acid
Isonicotinic acid
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 315…319 °C
Storage Temperature: Ambient
MDL Number: MFCD00006429
CAS Number: 55-22-1
EINECS: 200-228-2
Merck Index: 13,05206

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR