1-Hexanesulfonic acid sodium salt

Synonyms: Sodium hexane-1-sulphonate, Sodium hexanesulphonate, Sodium 1-hexanesulphonate

H9026-5G H9026-25G
220182-131EA 159.71 USD
220182-131 220182-130
1-Hexanesulfonic acid sodium salt
1-Hexanesulphonic acid sodium salt
Formula: C₆H₁₃O₃SNa
MW: 188.22 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007542
CAS Number: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=d608216fe1e698d6b206b03588e6d6fe5ee661e1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR