5-Hydroxyindol-3-ylacetic acid

Synonyms: 5-Hydroxyindole-3-acetic acid, 2-(5-Hydroxy-1H-indol-3-yl)acetic acid

H8876-1G H8876-100MG H8876-500MG
220182-750EA 232.89 USD
220182-750 220182-749 220182-751
5-Hydroxyindol-3-ylacetic acid
5-Hydroxyindol-3-ylacetic acid
Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Melting Pt: 166 °C
Storage Temperature: Freezer
MDL Number: MFCD00005639
CAS Number: 54-16-0
EINECS: 200-195-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR