L(+)-Histidine monohydrochloride monohydrate
Synonyms:
L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate
H8125-100G
H8125-5G
H8125-25G
H8125-5KG
H8125-1KG
220182-600EA
122.71
USD
220182-600
220182-603
220182-602
220182-604
220182-601
L(+)-Histidine monohydrochloride monohydrate
L(+)-Histidine monohydrochloride monohydrate
Formula:
C₆H₉N₃O₂·HCl·H₂O MW: 209.63 g/mol Melting Pt: 259 °C Density: 1.485 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151027 CAS Number: 5934-29-2 EINECS: 211-438-9 Merck Index: 13,04741 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...