cis-4-Hydroxy-D(+)-proline

Synonyms: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline

H5877-250MG H5877-10G H5877-1G
220182-355EA 42.34 USD
220182-355 220182-353 220182-354
cis-4-Hydroxy-D(+)-proline
cis-4-Hydroxy-D(+)-proline
Formula: C₅H₉NO₃
MW: 131.13 g/mol
MDL Number: MFCD00005252
CAS Number: 2584-71-6
EINECS: 219-963-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR