3-Pyridone

Warning

Synonyms: 3-Hydroxypyridine, β-Pyridone, Pyridin-3-ol, 3-Pyridinol

H57009-100G H57009-500G H57009-25G H57009-5KG
220182-334EA 47.92 USD
220182-334 220182-336 220182-335 220182-337
3-Pyridone
3-Pyridone
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 124…126 °C
Density: 1.2 g/cm³ (-70 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006378
CAS Number: 109-00-2
EINECS: 203-637-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR