trans-4-Hydroxy-L(-)-proline

Synonyms: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH

H5534-10MG H5534-25G H5534-1KG H5534-100G
220182-318EA 49.35 USD
220182-318 220182-320 220182-319 220182-317
trans-4-Hydroxy-L(-)-proline
trans-4-Hydroxy-L(-)-proline
Formula: C₅H₉NO₃
MW: 131.13 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00064320
CAS Number: 51-35-4
EINECS: 200-091-9
Merck Index: 13,04866

Order Now

Specification Test Results

Assay 98.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR