Tris(perfluorobutyl)amine

Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

H5262-25ML H5262-5ML
220182-272EA 394.62 USD
220182-272 220182-273
Tris(perfluorobutyl)amine
Tris(perfluorobutyl)amine
Formula: C₁₂F₂₇N
MW: 671.1 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –55 °C
Density: 1.883 g/cm³ (20 °C)
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR