3-(4-Hydroxyphenyl)propionic acid ≥98%

Synonyms: 3-(4-Hydroxyphenyl)propanoic acid, p-hydroxyl phenylpropanol, 3-(p-Hydroxyphenyl)-Propionic Acid

H52406-5G H52406-25G H52406-50G
220182-268EA 31.98 USD
220182-268 220182-266 220182-267
3-(4-Hydroxyphenyl)propionic acid ≥98%
3-(4-Hydroxyphenyl)propionic acid
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 208…210 °C (14 mmHg)
Melting Pt: 126…129 °C
Density: 1.336 g/cm³ (20 °C)
MDL Number: MFCD00002778
CAS Number: 501-97-3
EINECS: 207-931-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR