2-Hydroxy-5-nitropyridine

Synonyms: 5-Nitro-2-pyridinol

H48808-25G H48808-100G
220182-244EA 82.71 USD
220182-244 220182-243
2-Hydroxy-5-nitropyridine
2-Hydroxy-5-nitropyridine
Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Boiling Pt: 180 - 200 °C (0.2 -rr)
Melting Pt: 191 - 192 °C
MDL Number: MFCD00006276
CAS Number: 5418-51-9
EINECS: 226-525-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR