3-Hydroxy-4-nitrobenzoic acid ≥95%

Warning

H48409-25G H48409-100G
220182-242EA 131.38 USD
220182-242 220182-241
3-Hydroxy-4-nitrobenzoic acid ≥95%
3-Hydroxy-4-nitrobenzoic acid
Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 234 - 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00007110
CAS Number: 619-14-7
EINECS: 210-580-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR