5-Hydroxy-2-nitrobenzaldehyde ≥98%

Warning

H48107-25G H48107-5G
220182-239EA 157.23 USD
220182-239 220182-240
5-Hydroxy-2-nitrobenzaldehyde ≥98%
5-Hydroxy-2-nitrobenzaldehyde
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007332
CAS Number: 42454-06-8
EINECS: 255-832-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR