2-Hydroxy-1-naphthoic acid

H45809-250G H45809-5G
220182-206EA 116.31 USD
220182-206 220182-207
2-Hydroxy-1-naphthoic acid
2-Hydroxy-1-naphthoic acid
Formula: C₁₁H₈O₃
MW: 188.18 g/mol
MDL Number: MFCD00004009
CAS Number: 2283-08-1
EINECS: 218-919-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR