2-Methyl-4-quinolinol ≥98.5%

Warning

Synonyms: 4-Hydroxy-2-methylquinoline

H43806-50G H43806-10G
220182-159EA 320.92 USD
220182-159 220182-158
2-Methyl-4-quinolinol ≥98.5%
2-Methyl-4-quinolinol
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 313 °C (1013 hPa)
MDL Number: MFCD00006758
CAS Number: 607-67-0
EINECS: 210-140-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR