4-Hydroxy-4-methyl-2-pentanone ≥99%

Danger

Synonyms: Diacetone alcohol

H41544-25ML H41544-500ML H41544-4L
220182-074EA 35.22 USD
220182-074 220182-076 220182-075
4-Hydroxy-4-methyl-2-pentanone ≥99%
4-Hydroxy-4-methyl-2-pentanone
Formula: (CH₃)₂C(OH)CH₂COCH₃
MW: 116.16 g/mol
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
ADR: 3,III
REACH: 01-2119473975-21
Merck Index: 13,02983
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2da78bff918d252787ee81ab8857bd3b19e51ac&vid=d8a0cccff2cac15b826ba733a013451ac11e3b51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR