5-(Hydroxymethyl)-2-furaldehyde

Warning

Synonyms: 5-Hydroxymethyl-2-furancarbaldehyde, 5-(hydroxymethyl)furan-2-carbaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethylfurfural

H40807-1G H40807-250MG H40807-5G
220179-945EA 80.12 USD
220179-945 220179-946 220179-947
5-(Hydroxymethyl)-2-furaldehyde
5-(Hydroxymethyl)-2-furaldehyde
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 27…30 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003234
CAS Number: 67-47-0
EINECS: 200-654-9
Merck Index: 13,04857

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR