Hydrocortisone

H4001-100G H4001-10G H4001-25G H4001-5G H4001-1G
220182-059EA 1659.69 USD
220182-059 220182-060 220182-062 220182-063 220182-061
Hydrocortisone
Hydrocortisone
Formula: C₂₁H₃₀O₅
MW: 362.47 g/mol
Boiling Pt: 567 °C (1013 hPa)
Melting Pt: 202 - 204 °C
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00011654
CAS Number: 50-23-7
EINECS: 200-020-1

Order Now

Specification Test Results

Assay 98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR