3-Hydroxy-3-methylbutanone ≥95%

Synonyms: 3-Hydroxy-3-methyl-2-butanone, 3-Hydroxy-3-methylbutan-2-one

H39604-25G H39604-5G
220182-056EA 569.97 USD
220182-056 220182-057
3-Hydroxy-3-methylbutanone ≥95%
3-Hydroxy-3-methylbutanone
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 140 °C (1013 hPa)
Density: 0.9595 g/cm³ (13 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004460
CAS Number: 115-22-0
EINECS: 204-073-1
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR