Oxybenzone ≥98%

Synonyms: Eusolex® 4360, 2-Hydroxy-4-methoxybenzophenone, (2-Hydroxy-4-methoxyphenyl)(phenyl)methanone

H36206-100G H36206-5G H36206-500G
220181-173EA 91.22 USD
220181-173 220181-175 220181-174
Oxybenzone ≥98%
Oxybenzone
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Boiling Pt: 370 °C (1013 hPa)
Melting Pt: 65 °C
Density: 1.3397 g/cm³ (25 °C)
MDL Number: MFCD00008387
CAS Number: 131-57-7
EINECS: 205-031-5
Merck Index: 12,07088

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR