Patton-Reeders reagent (Calconcarboxylic acid) dihydrate

Warning

H3128-25G H3128-5G H3128-100G
220180-659EA 204.4 USD
220180-659 220180-660 220180-658
Patton-Reeders reagent (Calconcarboxylic acid) dihydrate
Patton-reeders reagent (Calconcarboxylic acid dihydrate)
Formula: C₂₁H₁₄N₂O₇S·2H₂O
MW: 474.45 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004078
CAS Number: 3737-95-9
EINECS: 223-117-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR