(±)-2-Heptanol ≥98%

Synonyms: (±)-Methyl pentyl carbinol, (±)-2-Hydroxy heptane, Heptan-2-ol, (±)-Pentyl methyl carbinol

H3003-100G-A H3003-25G-A
220181-777EA 70 USD
220181-777 220181-778
(±)-2-Heptanol ≥98%
(±)-2-Heptanol
Formula: C₇H₁₆O
MW: 116.2 g/mol
Boiling Pt: 160 °C (1013 hPa)
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004587
CAS Number: 543-49-7
EINECS: 208-844-3
UN: 1987
ADR: 3,III
Merck Index: 13,04682

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR