1-(2-Hydroxyethyl)piperazine

Danger

Synonyms: N-(2-Hydroxyethyl)piperazine, 2-(1-Piperazinyl)ethanol, 2-Piperazin-1-ylethanol

H28807-500G H28807-100G
220181-759EA 151.85 USD
220181-759 220181-758
1-(2-Hydroxyethyl)piperazine
1-(2-Hydroxyethyl)piperazine
Formula: C₆H₁₄N₂O
MW: 130.19 g/mol
Boiling Pt: ∼246 °C (1013 hPa)
Melting Pt: –38 °C
Density: 1.0595 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005970
CAS Number: 103-76-4
EINECS: 203-142-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR