1-Heptanol ≥98%

Warning

Synonyms: Heptyl alcohol

H2805-1L H2805-2.5L H2805-250ML
220181-752EA 133.54 USD
220181-752 220181-753 220181-754
1-Heptanol ≥98%
1-Heptanol
Formula: H₃C(CH₂)₅CH₂OH
MW: 116.2 g/mol
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002986
CAS Number: 111-70-6
EINECS: 203-897-9
Merck Index: 13,04681

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR