4-Hydroxycoumarin

Synonyms: 4-Hydroxy-2-benzopyrone

H23805-500G H23805-100G
220180-634EA 205.69 USD
220180-634 220180-633
4-Hydroxycoumarin
4-Hydroxycoumarin
Formula: C₉H₆O₃
MW: 162.14 g/mol
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 211 - 214 °C
MDL Number: MFCD00006856
CAS Number: 1076-38-6
EINECS: 214-060-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR