trans-2-Hydroxycinnamic acid predominantly trans

Warning

Synonyms: (E)-3-(2-Hydroxyphenyl)acrylic acid, 2-Hydroxycinnamic acid, predominantly trans, trans-2-Hydroxycinnamic acid, trans-o-Coumaric acid, o-Coumaric acid

H22809-5G H22809-25G
220179-528EA 91.54 USD
220179-528 220180-631
trans-2-Hydroxycinnamic acid predominantly trans
trans-2-Hydroxycinnamic acid
Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 217 °C
MDL Number: MFCD00004379
CAS Number: 614-60-8
EINECS: 210-386-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR