(4-Hydroxyphenyl)acetonitrile ≥98%

Synonyms: 4-Hydroxybenzyl cyanide, 2-(4-Hydroxyphenyl)acetonitrile

H21101-25G H21101-5G
220178-706EA 40.49 USD
220178-706 220178-707
(4-Hydroxyphenyl)acetonitrile ≥98%
(4-Hydroxyphenyl)acetonitrile
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 68…71 °C
MDL Number: MFCD00002383
CAS Number: 14191-95-8
EINECS: 238-046-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR