4-Hydroxybenzyl alcohol ≥99%

Synonyms: (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol

H20806-10G H20806-25G H20806-100G
220178-704EA 60.95 USD
220178-704 220178-705 220178-703
4-Hydroxybenzyl alcohol ≥99%
4-Hydroxybenzyl alcohol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: 115…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004658
CAS Number: 623-05-2
EINECS: 210-768-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR