3-Hydroxybenzyl alcohol ≥99%

Synonyms: (3-Hydroxyphenyl)methanol, KSD 2405, 3-(Hydroxymethyl)phenol

H20601-5G H20601-25G
220178-702EA 64.62 USD
220178-702 220178-701
3-Hydroxybenzyl alcohol ≥99%
3-Hydroxybenzyl alcohol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 86…87 °C (0.001 mmHg)
Melting Pt: 70 °C
Density: 1.161 g/cm³ (25 °C)
MDL Number: MFCD00004643
CAS Number: 620-24-6
EINECS: 210-633-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR