4-Hydroxybenzoic acid ≥99%

Warning

Synonyms: p-Hydroxybenzoic Acid

H20059-1KG H20059-500G H20059-100G
220178-690EA 105.65 USD
220178-690 220178-691 220178-689
4-Hydroxybenzoic acid ≥99%
4-Hydroxybenzoic acid
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 210 - 211 °C
Density: 1.398 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002547
CAS Number: 99-96-7
EINECS: 202-804-9
Merck Index: 13,04837

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR