3-Hydroxybenzoic acid ≥99%

Warning

Synonyms: m-Hydroxybenzoic Acid

H20008-500G H20008-5G H20008-100G
220181-664EA 229.38 USD
220181-664 220178-684 220181-663
3-Hydroxybenzoic acid ≥99%
3-Hydroxybenzoic acid
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 346 °C (1013 hPa)
Melting Pt: 199 °C
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002506
CAS Number: 99-06-9
EINECS: 202-726-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR