3-Hydroxybenzaldehyde ≥97%

Warning

H19808-5G H19808-500G H19808-100G
220181-660EA 36.62 USD
220181-660 220181-659 220181-658
3-Hydroxybenzaldehyde ≥97%
3-Hydroxybenzaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 99…102 °C
Density: 1.1299 g/cm³ (114 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003368
CAS Number: 100-83-4
EINECS: 202-892-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR