2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%

Danger

Synonyms: 1H,1H-Perfluoro-1-butanol, 2,2,3,3,4,4,4-Heptafluorobutan-1-ol, Heptafluoro-1-butanol

H1604-5G H1604-25G
220180-602EA 49.65 USD
220180-602 220180-601
2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%
2,2,3,3,4,4,4-Heptafluoro-1-butanol
Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 96 °C (1013 hPa)
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004674
CAS Number: 375-01-9
EINECS: 206-782-1
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR