n-Hexatriacontane ≥98%

Synonyms: Hexatriacontane

H12552-50G H12552-25G H12552-5G
220181-539EA 214.31 USD
220181-539 220181-538 220181-540
n-Hexatriacontane ≥98%
n-Hexatriacontane
Formula: H₃C(CH₂)₃₄CH₃
MW: 506.98 g/mol
Boiling Pt: 497 °C (1013 hPa)
Melting Pt: 75.5 °C
Density: 0.7803 g/cm³ (80 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009413
CAS Number: 630-06-8
EINECS: 211-127-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR