(±)-3-Hexanol ≥97%

Synonyms: (±)-Propyl ethyl carbinol, (±)-Ethyl propyl carbinol, (±)-3-Hydroxy hexane

H12404-25G H12404-100G
220181-529EA 56.63 USD
220181-529 220181-528
(±)-3-Hexanol ≥97%
(±)-3-Hexanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: -70 °C
Density: 0.819 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004582
CAS Number: 623-37-0
EINECS: 210-790-0
UN: 2282
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39530c9bef58d2cdcee4bbc14b525d77fdf89c21&vid=f007f095722cbdace163e1263edb2bf7a2739678&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR