1,6-Hexanediol ≥97%

Synonyms: Hexamethylene glycol, 1,6-Dihydroxy hexane

H11807-500G H11807-1KG
220181-520EA 84.65 USD
220181-520 220181-519
1,6-Hexanediol ≥97%
1,6-Hexanediol
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 39…42 °C
Density: 1.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002985
CAS Number: 629-11-8
EINECS: 211-074-0
Merck Index: 13,04709

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR