1,6-Diaminohexane ≥98%

Danger

Synonyms: 1,6-Hexanediamine, Hexamethylenediamine

H11696-1KG H11696-500G H11696-100G H11696-25G
220181-516EA 142.15 USD
220181-516 220181-518 220181-515 220181-517
1,6-Diaminohexane ≥98%
1,6-Diaminohexane
Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 39…42 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00008243
CAS Number: 124-09-4
EINECS: 204-679-6
UN: 2280
ADR: 8,III
Merck Index: 13,04713

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR