(±)-4-Hydroxy-3-methoxymandelic acid ≥98%

Warning

Synonyms: 4-Hydroxy-3-methoxy-DL-mandelic acid, VMA, Vanillyl mandelic acid, DL-4-Hydroxy-3-methoxymandelic acid, (±)-4-Hydroxy-3-methoxyphenylglycolic acid, 2-Hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid, Vanillylmandelic acid

H0131-250MG H0131-500MG H0131-100MG H0131-5G H0131-1G
220180-573EA 75.35 USD
220180-573 220180-574 220180-571 220180-575 220180-572
(±)-4-Hydroxy-3-methoxymandelic acid ≥98%
(±)-4-Hydroxy-3-methoxymandelic acid
Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Boiling Pt: 421 °C (1013 hPa)
Melting Pt: 132 - 134 °C
Density: 1.44 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004235
CAS Number: 55-10-7
EINECS: 200-224-0
Merck Index: 12,10070

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR