Gallic acid

Synonyms: 3,4,5-Trihydroxybenzoic acid

G7384-1KG G7384-100G G7384-250G
220180-144EA 527.31 USD
220180-144 220180-143 220180-145
Gallic acid
Gallic acid
Formula: C₇H₆O₅
MW: 170.12 g/mol
Boiling Pt: 501 °C (1013 hPa)
Melting Pt: 251…252 °C
Density: 1.694 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002510
CAS Number: 149-91-7
EINECS: 205-749-9
Merck Index: 13,04366

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR