Guanidinium thiocyanate

Warning

G6639-250G G6639-100G G6639-500G G6639-1KG G6639-25G
220179-976EA 327.22 USD
220179-976 220179-974 220179-978 220179-975 220179-977
Guanidinium thiocyanate
Guanidinium thiocyanate
Formula: CH₅N₃·CHNS
Melting Pt: 118 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR