Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) ≥99%, Sigma-Aldrich®

Synonyms: H-Gly-OEt.HCl, Glycine Ethyl Ester HCl, Ethyl Glycinate Hydrochloride

M-Clarity™ quality level = MQ200

G6503-5G-A G6503-5G G6503 G6503-500G-A G6503-100G-A
220178-193EA 36.62 USD
220178-193 220178-192 220178-190 220178-191 220178-189
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) ≥99%, Sigma-Aldrich®
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl)
Formula: C₄H₁₀ClNO₂
MW: 139.58 g/mol
MDL Number: MFCD00012871
CAS Number: 623-33-6
EINECS: 207-331-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR