Giemsa's stain, certified by the Biological Stain Commission, Sigma-Aldrich®

Warning

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

G5637-5G G5637-25G
220179-731EA 156.8 USD
220179-731 220179-730
Giemsa's stain, certified by the Biological Stain Commission, Sigma-Aldrich®
Giemsa's stain

Certified for Giemsa stain for blood films and for the Wolbach′s Giemsa method for paraffin sections

Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
EINECS: 257-438-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=350eadceb48bdd9140015036a7711c27eda3ceed&vid=4cff803bb4f00f2a35563329e7d579458ee98aa5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR