L(-)-Glucose

Synonyms: (2S,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal

G5500-500MG G5500-1G G5500-250MG G5500-25G G5500-5G
220179-693EA 116.06 USD
220179-693 220179-690 220179-691 220179-692 220179-694
L(-)-Glucose
L(-)-Glucose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 146…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00148913
CAS Number: 921-60-8
EINECS: 213-068-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR