D-Glucono-1,5-lactone

Synonyms: GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone

G4750-1KG G4750-100G G4750-500G
220179-322EA 252.31 USD
220179-322 220179-321 220179-323
D-Glucono-1,5-lactone
D-Glucono-1,5-lactone
Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 385 °C (1013 hPa)
Melting Pt: 153 °C
Density: 1.61 g/cm³ (-5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006647
CAS Number: 90-80-2
EINECS: 202-016-5
Merck Index: 13,04470

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR