Calcium-D-gluconate monohydrate ≥98%

G4625-1KG G4625-500G
220179-301EA 141.54 USD
220179-301 220179-302
Calcium-D-gluconate monohydrate ≥98%
Calcium-D-gluconate monohydrate
Formula: C₁₂H₂₂CaO₁₄·H₂O
MW: 448.39 g/mol
Melting Pt: 178 °C
MDL Number: MFCD00150787
CAS Number: 66905-23-5
EINECS: 206-075-8
Merck Index: 13,01671

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR