L(-)-Glutathione (reduced form) ≥98.0%

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

G4251-100G G4251-500G G4251-250G G4251-5G G4251-10G G4251-1G G4251-300MG G4251-50G G4251-10MG G4251-25G
220179-237EA 733.54 USD
220179-237 220179-244 220179-241 220179-246 220179-238 220179-240 220179-243 220179-245 220179-239 220179-242
L(-)-Glutathione (reduced form) ≥98.0%
L(-)-Glutathione (reduced form)
Formula: C₁₀H₁₇N₃O₆S
MW: 307.33 g/mol
Melting Pt: 185…195 °C
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488

Order Now

Specification Test Results

Assay 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR