tetra-Chloroauric(III) acid trihydrate ACS

Danger

Synonyms: Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate

G4022-10G G4022-25G G4022-1G G4022-5G
220179-193EA 605.54 USD
220179-193 220179-195 220179-194 220179-196
tetra-Chloroauric(III) acid trihydrate ACS
tetra-Chloroauric(III) acid trihydrate
Formula: HAuCl₄·3H₂O
MW: 393.83 g/mol
Melting Pt: 30 °C
Density: 3.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149904
CAS Number: 16961-25-4
EINECS: 240-948-4
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR